2-Cyano-3-methylpyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019458-25G 019458-100G
FLUO019458-25GEA 0 GBP
FLUO019458-25G FLUO019458-100G
2-Cyano-3-methylpyridine
2-Cyano-3-methylpyridine
Formula: C₇H₆N₂
Boiling Pt: 139…142 °C (38 mmHg)
Melting Pt: 87…90 °C
MDL Number: MFCD00190585
CAS Number: 20970-75-6
EINECS: 244-131-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR