2-Chloro-6-nitrobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CA-4251-25G CA-4251-100G CA-4251-5G
COBBCA-4251-25GEA 0 GBP
COBBCA-4251-25G COBBCA-4251-100G COBBCA-4251-5G
2-Chloro-6-nitrobenzoic acid
2-Chloro-6-nitrobenzoic acid
Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
MDL Number: MFCD00100425
CAS Number: 5344-49-0
EINECS: 226-287-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR