2-Chloro-6-methoxypyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
OS-7654-5G
OS-7654-25G
OS-7654-100G
COBBOS-7654-5GEA
0
GBP
COBBOS-7654-5G
COBBOS-7654-25G
COBBOS-7654-100G
2-Chloro-6-methoxypyridine
2-Chloro-6-methoxypyridine
Formula:
C₆H₆ClNO MW: 143.57 g/mol Boiling Pt: 39…40 °C (0.3 mmHg) Density: 1.207 g/cm³ (20 °C) |
MDL Number:
MFCD00006265 CAS Number: 17228-64-7 EINECS: 241-264-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...