2-Chloro-6-fluorophenylboronic acid ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
(2-Chloro-6-fluorophenyl)boronic acid, (2-Chloro-6-fluorophenyl)metaboric acid, 2-Chloro-6-fluorobenzeneboronic acid, Dihydroxy(2-chloro-6-fluorophenyl)borane
H53079.03
H53079.06
H53079.03EA
0
GBP
H53079.03
H53079.06
2-Chloro-6-fluorophenylboronic acid ≥97%
2-Chloro-6-fluorophenylboronic acid
Purity: 97%
CAS No.: 313545-32-3
Molecular Formula: C6H5BClFO2
Formula Weight: 174.37
Melting Point: 131-135°
MDL No.: MFCD04039892
Formula:
C₆H₅BClFO₂ MW: 174.37 g/mol Melting Pt: 131…135 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039892 CAS Number: 313545-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...