2-Chloro-6-(trifluoromethyl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1127-100G PY-1127-25G PY-1127-5G
COBBPY-1127-100GEA 0 GBP
COBBPY-1127-100G COBBPY-1127-25G COBBPY-1127-5G
2-Chloro-6-(trifluoromethyl)pyridine
2-Chloro-6-(trifluoromethyl)pyridine
Formula: C₆H₃ClF₃N
MDL Number: MFCD00042226
CAS Number: 39890-95-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR