Methyl 2-chloro-5-fluorobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4571-1G CA-4571-25G CA-4571-5G
COBBCA-4571-1GEA 0 GBP
COBBCA-4571-1G COBBCA-4571-25G COBBCA-4571-5G
Methyl 2-chloro-5-fluorobenzoate
Methyl 2-chloro-5-fluorobenzoate
Formula: C₈H₆ClFO₂
MDL Number: MFCD06204207
CAS Number: 647020-63-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR