2-Chloro-4-pyridineboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(2-Chloropyridin-4-yl)boronic acid, 2-Chloro-4-pyridinylboronic acid, 2-Chloropyridine-4-boronic acid
BB-2272-1G
BB-2272-5G
COBBBB-2272-1GEA
0
GBP
COBBBB-2272-1G
COBBBB-2272-5G
2-Chloro-4-pyridineboronic acid
2-Chloro-4-pyridineboronic acid
Formula:
C₅H₅BClNO₂ MW: 157.36 g/mol |
MDL Number:
MFCD03788416 CAS Number: 458532-96-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...