2-Chloro-4-methyl-5-nitropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7688-5G OS-7688-25G
COBBOS-7688-5GEA 0 GBP
COBBOS-7688-5G COBBOS-7688-25G
2-Chloro-4-methyl-5-nitropyridine
2-Chloro-4-methyl-5-nitropyridine
Formula: C₆H₅ClN₂O₂
MDL Number: MFCD00010688
CAS Number: 23056-33-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR