2-Chloro-4-fluorobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4386-100G CA-4386-25G
COBBCA-4386-100GEA 0 GBP
COBBCA-4386-100G COBBCA-4386-25G
2-Chloro-4-fluorobenzoic acid
2-Chloro-4-fluorobenzoic acid
Formula: C₇H₄ClFO₂
MW: 174.56 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00010615
CAS Number: 2252-51-9
EINECS: 218-845-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR