2-Chloro-3-hydroxybenzaldehyde, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

647365-10G 647365-1G
SIAL647365-10GEA 186 GBP
SIAL647365-10G SIAL647365-1G
2-Chloro-3-hydroxybenzaldehyde, Sigma-Aldrich®
2-Chloro-3-hydroxybenzaldehyde
Formula: C₇H₅ClO₂
MW: 156.57 g/mol
MDL Number: MFCD06798118
CAS Number: 56962-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR