2-Chloro-3-(trifluoromethyl)benzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-3-trifluoromethylbenzoic acid, 2-Chloro-α,α,α-trifluoro-m-toluic acid

OS-7466-100G OS-7466-5G OS-7466-1G OS-7466-25G
COBBOS-7466-100GEA 0 GBP
COBBOS-7466-100G COBBOS-7466-5G COBBOS-7466-1G COBBOS-7466-25G
2-Chloro-3-(trifluoromethyl)benzoic acid
2-Chloro-3-(trifluoromethyl)benzoic acid
Formula: C₈H₄ClF₃O₂
MW: 224.57 g/mol
MDL Number: MFCD00792415
CAS Number: 39226-97-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR