2-Bromothiobenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021313-1G 021313-5G
FLUO021313-1GEA 0 GBP
FLUO021313-1G FLUO021313-5G
2-Bromothiobenzamide
2-Bromothiobenzamide
Formula: C₇H₆BrNS
MDL Number: MFCD04973314
CAS Number: 30216-44-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR