2-Bromophenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2654-5G BB-2654-25G BB-2654-1G
COBBBB-2654-5GEA 0 GBP
COBBBB-2654-5G COBBBB-2654-25G COBBBB-2654-1G
2-Bromophenylboronic acid
2-Bromophenylboronic acid
Formula: C₆H₆BBrO₂
MDL Number: MFCD01114672
CAS Number: 244205-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR