2-Bromo-6-chlorobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OS-7468-5G OS-7468-25G
COBBOS-7468-5GEA 0 GBP
COBBOS-7468-5G COBBOS-7468-25G
2-Bromo-6-chlorobenzoic acid
2-Bromo-6-chlorobenzoic acid
Formula: C₇H₄BrClO₂
MW: 235.46 g/mol
MDL Number: MFCD00672929
CAS Number: 93224-85-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR