2-Bromo-5-methylthiophene

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Methyl-2-bromothiophene, 2-Bromo-5-methylthiophenol

HI-1254-25G HI-1254-100G HI-1254-5G
COBBHI-1254-25GEA 0 GBP
COBBHI-1254-25G COBBHI-1254-100G COBBHI-1254-5G
2-Bromo-5-methylthiophene
2-Bromo-5-methylthiophene
Formula: C₅H₅BrS
MW: 177.06 g/mol
Boiling Pt: 178
Melting Pt: 58
Density: 1.5529 g/cm³ (20 °C)
MDL Number: MFCD00130103
CAS Number: 765-58-2
EINECS: 212-151-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR