2-Bromo-5-methylpyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

OT-0356-25G OT-0356-5G OT-0356-100G
COBBOT-0356-25GEA 0 GBP
COBBOT-0356-25G COBBOT-0356-5G COBBOT-0356-100G
2-Bromo-5-methylpyridine
2-Bromo-5-methylpyridine
Formula: C₆H₆BrN
Boiling Pt: 65 °C (0,5 mmHg)
Melting Pt: 152,5 °C
MDL Number: MFCD00209553
CAS Number: 3510-66-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR