6-Bromo-m-toluic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7703-5G
OS-7703-25G
OS-7703-1G
COBBOS-7703-5GEA
0
GBP
COBBOS-7703-5G
COBBOS-7703-25G
COBBOS-7703-1G
6-Bromo-m-toluic acid
6-Bromo-m-toluic acid
Formula:
C₈H₇BrO₂ MW: 215.05 g/mol |
MDL Number:
MFCD00079722 CAS Number: 6967-82-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...