2-Bromo-5-methoxybenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4278-5G CA-4278-25G CA-4278-100G
COBBCA-4278-5GEA 0 GBP
COBBCA-4278-5G COBBCA-4278-25G COBBCA-4278-100G
2-Bromo-5-methoxybenzoic acid
2-Bromo-5-methoxybenzoic acid
Formula: C₈H₇BrO₃
MW: 231.05 g/mol
MDL Number: MFCD00020214
CAS Number: 22921-68-2
EINECS: 245-329-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR