2-Bromo-5-hydroxypyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Bromo-5-pyridinol, 6-Bromopyridin-3-ol
PY-7274-1G
PY-7274-25G
PY-7274-5G
COBBPY-7274-1GEA
0
GBP
COBBPY-7274-1G
COBBPY-7274-25G
COBBPY-7274-5G
2-Bromo-5-hydroxypyridine
2-Bromo-5-hydroxypyridine
Formula:
C₅H₄BrNO MW: 174 g/mol |
MDL Number:
MFCD04114184 CAS Number: 55717-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...