2-Bromo-5-fluorobenzoic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
CA-4003-25G
CA-4003-100G
COBBCA-4003-25GEA
0
GBP
COBBCA-4003-25G
COBBCA-4003-100G
2-Bromo-5-fluorobenzoic acid
2-Bromo-5-fluorobenzoic acid
Formula:
C₇H₄BrFO₂ MW: 219.01 g/mol |
MDL Number:
MFCD00142874 CAS Number: 394-28-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...