2-Bromo-5-chloro-3-nitropyridine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

024451-1G 024451-5G
FLUO024451-1GEA 0 GBP
FLUO024451-1G FLUO024451-5G
2-Bromo-5-chloro-3-nitropyridine
2-Bromo-5-chloro-3-nitropyridine
Formula: C₅H₂BrClN₂O₂
MDL Number: MFCD07368982
CAS Number: 75806-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR