2-Bromo-5-(trifluoromethyl)benzaldehyde

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-8059-5G OT-8059-25G OT-8059-1G
COBBOT-8059-5GEA 0 GBP
COBBOT-8059-5G COBBOT-8059-25G COBBOT-8059-1G
2-Bromo-5-(trifluoromethyl)benzaldehyde
2-Bromo-5-(trifluoromethyl)benzaldehyde
Formula: C₈H₄BrF₃O
MDL Number: MFCD04973760
CAS Number: 102684-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR