Methyl 2-bromo-4-nitrobenzoate

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4572-5G CA-4572-25G CA-4572-1G
COBBCA-4572-5GEA 0 GBP
COBBCA-4572-5G COBBCA-4572-25G COBBCA-4572-1G
Methyl 2-bromo-4-nitrobenzoate
Methyl 2-bromo-4-nitrobenzoate
Formula: C₈H₆BrNO₄
MDL Number: MFCD07779271
CAS Number: 100959-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR