2-Bromo-4-nitrobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4384-5G CA-4384-25G CA-4384-1G
COBBCA-4384-5GEA 0 GBP
COBBCA-4384-5G COBBCA-4384-25G COBBCA-4384-1G
2-Bromo-4-nitrobenzoic acid
2-Bromo-4-nitrobenzoic acid
Formula: C₇H₄BrNO₄
MDL Number: MFCD00234252
CAS Number: 16426-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR