2-Bromo-4-fluorobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CA-4002-5G CA-4002-25G CA-4002-100G
COBBCA-4002-5GEA 0 GBP
COBBCA-4002-5G COBBCA-4002-25G COBBCA-4002-100G
2-Bromo-4-fluorobenzoic acid
2-Bromo-4-fluorobenzoic acid
Formula: C₇H₄BrFO₂
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: 173…175 °C
MDL Number: MFCD00055370
CAS Number: 1006-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR