2-Bromoisonicotinonitrile

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7156-1G PY-7156-5G
COBBPY-7156-1GEA 0 GBP
COBBPY-7156-1G COBBPY-7156-5G
2-Bromoisonicotinonitrile
2-Bromoisonicotinonitrile
Formula: C₆H₃BrN₂
MW: 183.01 g/mol
MDL Number: MFCD07367879
CAS Number: 10386-27-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR