2-Bromo-3-methylthiophene
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
CS-9093-5G
CS-9093-25G
CS-9093-100G
COBBCS-9093-5GEA
0
GBP
COBBCS-9093-5G
COBBCS-9093-25G
COBBCS-9093-100G
2-Bromo-3-methylthiophene
2-Bromo-3-methylthiophene
Formula:
C₅H₅BrS MW: 177.06 g/mol Boiling Pt: 14…16 °C (0.3 mmHg) Density: 1.5844 g/cm³ (17.5 °C) Storage Temperature: Refrigerator |
MDL Number:
MFCD00059741 CAS Number: 14282-76-9 EINECS: 238-175-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...