2-Bromo-3-hydroxypyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2-Bromo-3-pyridinol
OS-7430-5G
OS-7430-25G
OS-7430-100G
COBBOS-7430-5GEA
0
GBP
COBBOS-7430-5G
COBBOS-7430-25G
COBBOS-7430-100G
2-Bromo-3-hydroxypyridine
2-Bromo-3-hydroxypyridine
Formula:
C₅H₄BrNO MW: 174 g/mol Boiling Pt: 232 Melting Pt: 185…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006220 CAS Number: 6602-32-0 EINECS: 229-547-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...