2-Aminoquinoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: Quinolin-2-ylamine

PY-7882-1G PY-7882-5G
COBBPY-7882-1GEA 0 GBP
COBBPY-7882-1G COBBPY-7882-5G
2-Aminoquinoline
2-Aminoquinoline
Formula: C₉H₈N₂
MW: 144.18 g/mol
Boiling Pt: 305 °C (1013 hPa)
Melting Pt: 130 °C
MDL Number: MFCD00038079
CAS Number: 580-22-3
EINECS: 209-458-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR