2-Amino-6-(trifluoromethyl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7905-1G PY-7905-5G
COBBPY-7905-1GEA 0 GBP
COBBPY-7905-1G COBBPY-7905-5G
2-Amino-6-(trifluoromethyl)pyridine
2-Amino-6-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
Boiling Pt: 200 °C (1013 hPa)
Melting Pt: 78…80 °C
MDL Number: MFCD03093868
CAS Number: 34486-24-3
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR