2-Amino-5-nitropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Nitropyridin-2-ylamine, 5-Nitropyridin-2-amine, 5-Nitro-2-pyridylamine

PY-7149-100G PY-7149-25G PY-7149-5G
COBBPY-7149-100GEA 0 GBP
COBBPY-7149-100G COBBPY-7149-25G COBBPY-7149-5G
2-Amino-5-nitropyridine
2-Amino-5-nitropyridine
Formula: C₅H₅N₃O₂
Melting Pt: 185…188 °C
Density: 1,623 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006325
CAS Number: 4214-76-0
EINECS: 224-145-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR