2-Amino-5-chloropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-7094-100G PY-7094-25G PY-7094-500G
COBBPY-7094-100GEA 0 GBP
COBBPY-7094-100G COBBPY-7094-25G COBBPY-7094-500G
2-Amino-5-chloropyridine
2-Amino-5-chloropyridine
Formula: C₅H₅ClN₂
Boiling Pt: 241 °C (1013 hPa)
Melting Pt: 129…131 °C
Density: 1,514 g/cm³ (20 °C)
MDL Number: MFCD00006324
CAS Number: 1072-98-6
EINECS: 214-020-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR