2-Amino-5-chloro-3-nitropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

PY-7763-25G PY-7763-5G
COBBPY-7763-25GEA 0 GBP
COBBPY-7763-25G COBBPY-7763-5G
2-Amino-5-chloro-3-nitropyridine
2-Amino-5-chloro-3-nitropyridine
Formula: C₅H₄ClN₃O₂
MDL Number: MFCD00092011
CAS Number: 5409-39-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR