2-Amino-5-bromopyrimidine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Bromopyrimidin-2-ylamine, 5-Bromopyrimidin-2-amine

OS-7511-25G OS-7511-5G
COBBOS-7511-25GEA 0 GBP
COBBOS-7511-25G COBBOS-7511-5G
2-Amino-5-bromopyrimidine
2-Amino-5-bromopyrimidine
Formula: C₄H₄BrN₃
MW: 174 g/mol
MDL Number: MFCD00012341
CAS Number: 7752-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR