2-Amino-5-bromopyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 5-Bromo-2-pyridylamine, 5-Bromopyridin-2-amine

OT-0326-25G OT-0326-100G
COBBOT-0326-25GEA 0 GBP
COBBOT-0326-25G COBBOT-0326-100G
2-Amino-5-bromopyridine
2-Amino-5-bromopyridine
Formula: C₅H₅BrN₂
MW: 173.01 g/mol
Boiling Pt: 244 °C (1013 hPa)
Melting Pt: 136…139 °C
Storage Temperature: Ambient
MDL Number: MFCD00006323
CAS Number: 1072-97-5
EINECS: 214-019-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR