6-Amino-3-bromo-2-methylpyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 6-Amino-3-bromo-2-picoline, 6-AMINO-3-BROMO-2-METHYLPYRIDINE, 3-Bromo-2-methyl-6-pyridylamine, 5-Bromo-6-methylpyridin-2-amine, 3-Bromo-2-methylpyridin-6-amine, 2-Amino-5-bromo-6-picoline, ['6-Amino-3-bromo-2-picoline'], 5-Bromo-6-methyl-2-pyridylamine, 2-Amino-5-bromo-6-methylpyridine

OT-0319-100G OT-0319-25G OT-0319-5G
COBBOT-0319-100GEA 0 GBP
COBBOT-0319-100G COBBOT-0319-25G COBBOT-0319-5G
6-Amino-3-bromo-2-methylpyridine
6-Amino-3-bromo-2-methylpyridine
Formula: C₆H₇BrN₂
MW: 187.04 g/mol
MDL Number: MFCD00068230
CAS Number: 42753-71-9
EINECS: 255-927-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR