2-Amino-5-nitro-4-picoline

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Methyl-5-nitro-2-pyridylamine, 4-Methyl-5-nitropyridin-2-amine, 2-Amino-4-methyl-5-nitropyridine

PY-7026-100G PY-7026-1G PY-7026-25G PY-7026-5G
COBBPY-7026-100GEA 0 GBP
COBBPY-7026-100G COBBPY-7026-1G COBBPY-7026-25G COBBPY-7026-5G
2-Amino-5-nitro-4-picoline
2-Amino-5-nitro-4-picoline
Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
Melting Pt: 220 °C
MDL Number: MFCD00010692
CAS Number: 21901-40-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR