2-Amino-4-methyl-3-nitropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Methyl-3-nitro-2-pyridylamine, 4-Methyl-3-nitropyridin-2-amine, 2-Amino-3-nitro-4-picoline

PY-7800-100G PY-7800-25G PY-7800-5G
COBBPY-7800-100GEA 0 GBP
COBBPY-7800-100G COBBPY-7800-25G COBBPY-7800-5G
2-Amino-4-methyl-3-nitropyridine
2-Amino-4-methyl-3-nitropyridine
Formula: C₆H₇N₃O₂
Melting Pt: 140 °C
MDL Number: MFCD00006315
CAS Number: 6635-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR