2-Amino-4-chloropyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Chloro-2-pyridylamine, 4-Chloropyridin-2-amine

OT-0380-5G OT-0380-25G OT-0380-100G
COBBOT-0380-5GEA 0 GBP
COBBOT-0380-5G COBBOT-0380-25G COBBOT-0380-100G
2-Amino-4-chloropyridine
2-Amino-4-chloropyridine
Formula: C₅H₅ClN₂
MW: 128.56 g/mol
MDL Number: MFCD04113820
CAS Number: 19798-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR