2-Amino-4-(trifluoromethyl)pyridine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

PY-1456-1G PY-1456-25G PY-1456-5G
COBBPY-1456-1GEA 0 GBP
COBBPY-1456-1G COBBPY-1456-25G COBBPY-1456-5G
2-Amino-4-(trifluoromethyl)pyridine
2-Amino-4-(trifluoromethyl)pyridine
Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
MDL Number: MFCD00077455
CAS Number: 106447-97-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR