2-Aminonicotinic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
2-Aminopyridine-3-carboxylic acid, 2-Amino-3-pyridinecarboxylic acid
CA-4056-100G
CA-4056-25G
COBBCA-4056-100GEA
0
GBP
COBBCA-4056-100G
COBBCA-4056-25G
2-Aminonicotinic acid
2-Aminonicotinic acid
Formula:
C₆H₆N₂O₂ MW: 138.13 g/mol Melting Pt: 297 Density: 1.545 g/cm³ (20 °C) |
MDL Number:
MFCD00006318 CAS Number: 5345-47-1 EINECS: 226-296-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...