2-Amino-3-bromo-5-fluoropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
PY-7239-1G
PY-7239-5G
COBBPY-7239-1GEA
0
GBP
COBBPY-7239-1G
COBBPY-7239-5G
2-Amino-3-bromo-5-fluoropyridine
2-Amino-3-bromo-5-fluoropyridine
Formula:
C₅H₄BrFN₂ |
MDL Number:
MFCD03092924 CAS Number: 869557-43-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...