2-Amino-3-bromo-5-chloropyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
3-Bromo-5-chloro-2-pyridinamine, 3-Bromo-5-chloro-2-pyridylamine, 3-Bromo-5-chloropyridin-2-amine
PY-7101-25G
PY-7101-5G
COBBPY-7101-25GEA
0
GBP
COBBPY-7101-25G
COBBPY-7101-5G
2-Amino-3-bromo-5-chloropyridine
2-Amino-3-bromo-5-chloropyridine
Formula:
C₅H₄BrClN₂ |
MDL Number:
MFCD00955627 CAS Number: 26163-03-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...