Pyridine-2,6-dicarboxylic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2,6-Pyridinedicarboxylic acid, Dipicolinic acid, DPA
PY-7340-100G
PY-7340-25G
COBBPY-7340-100GEA
0
GBP
COBBPY-7340-100G
COBBPY-7340-25G
Pyridine-2,6-dicarboxylic acid
Pyridine-2,6-dicarboxylic acid
Formula:
C₇H₅NO₄ Boiling Pt: 357 °C (1013 hPa) Melting Pt: 327 °C Flash Pt: 188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006299 CAS Number: 499-83-2 EINECS: 207-894-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...