2,6-Dimethylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,6-Dimethylbenzeneboronic acid, (2,6-Dimethylphenyl)boronic acid, Dihydroxy(2,6-dimethylphenyl)borane, (2,6-Dimethylphenyl)metaboric acid

BB-2205-5G BB-2205-25G BB-2205-1G
COBBBB-2205-5GEA 0 GBP
COBBBB-2205-5G COBBBB-2205-25G COBBBB-2205-1G
2,6-Dimethylphenylboronic acid
2,6-Dimethylphenylboronic acid
Formula: C₈H₁₁BO₂
MW: 149.98 g/mol
MDL Number: MFCD01009693
CAS Number: 100379-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR