2,6-Dimethoxypyridine
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
OS-7563-100G
OS-7563-25G
OS-7563-5G
COBBOS-7563-100GEA
0
GBP
COBBOS-7563-100G
COBBOS-7563-25G
COBBOS-7563-5G
2,6-Dimethoxypyridine
2,6-Dimethoxypyridine
Formula:
C₇H₉NO₂ MW: 139.15 g/mol Boiling Pt: 178 °C (1013 hPa) Density: 1,05 g/cm³ (20 °C) |
MDL Number:
MFCD00006266 CAS Number: 6231-18-1 EINECS: 228-334-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...