2,6-Dimethoxyphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2592-5G BB-2592-25G BB-2592-100G
COBBBB-2592-5GEA 0 GBP
COBBBB-2592-5G COBBBB-2592-25G COBBBB-2592-100G
2,6-Dimethoxyphenylboronic acid
2,6-Dimethoxyphenylboronic acid
Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
MDL Number: MFCD01318987
CAS Number: 23112-96-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR