2,6-Difluorobenzenesulphonamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CFF263.5 CFF263.10 CFF263.25 017310-5G 017310-25G 017310-1G
CHMPCFF263.5EA 0 GBP
CHMPCFF263.5 CHMPCFF263.10 CHMPCFF263.25 FLUO017310-5G FLUO017310-25G FLUO017310-1G
2,6-Difluorobenzenesulphonamide
2,6-Difluorobenzenesulphonamide
Formula: C₆H₅F₂NO₂S
MDL Number: MFCD00729101
CAS Number: 60230-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR