2,6-Difluorobenzoic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
CA-4073-100G
CA-4073-5G
CA-4073-25G
COBBCA-4073-100GEA
0
GBP
COBBCA-4073-100G
COBBCA-4073-5G
COBBCA-4073-25G
2,6-Difluorobenzoic acid
2,6-Difluorobenzoic acid
Formula:
C₇H₄F₂O₂ MW: 158.1 g/mol Boiling Pt: 72…77 °C (13 mmHg) Melting Pt: 159…160 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002411 CAS Number: 385-00-2 EINECS: 206-856-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...