2,6-Difluoro-3-nitrobenzoic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

CA-4078-5G CA-4078-25G CA-4078-1G
COBBCA-4078-5GEA 0 GBP
COBBCA-4078-5G COBBCA-4078-25G COBBCA-4078-1G
2,6-Difluoro-3-nitrobenzoic acid
2,6-Difluoro-3-nitrobenzoic acid
Formula: C₇H₃F₂NO₄
MDL Number: MFCD00119291
CAS Number: 83141-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR